Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099952.2 | NP_001093422.2 | 7, 8 | 882 | ||
NM_001168272.1 | NP_001161744.1 | 7, 8 | 882 | |||
NM_002222.5 | NP_002213.5 | 7, 8 | 882 | |||
XM_005265109.2 | XP_005265166.1 | 7, 8 | 882 | |||
XM_005265110.2 | XP_005265167.1 | 7, 8 | 882 | |||
XM_006713131.2 | XP_006713194.1 | 7, 8 | 882 | |||
XM_011533681.1 | XP_011531983.1 | 7, 8 | 882 | |||
XM_011533682.2 | XP_011531984.1 | 7, 8 | 808 | |||
XM_011533683.2 | XP_011531985.1 | 7, 8 | 882 | |||
XM_011533684.1 | XP_011531986.1 | 7, 8 | 882 | |||
XM_011533685.1 | XP_011531987.1 | 7, 8 | 882 | |||
XM_011533686.1 | XP_011531988.1 | 7, 8 | 882 | |||
XM_011533687.1 | XP_011531989.1 | 7, 8 | 882 | |||
XM_011533688.1 | XP_011531990.1 | 7, 8 | 882 | |||
XM_011533690.1 | XP_011531992.1 | 7, 8 | 882 | |||
XM_011533691.1 | XP_011531993.1 | 7, 8 | 882 | |||
XM_011533692.2 | XP_011531994.1 | 7, 8 | 882 | |||
XM_017006357.1 | XP_016861846.1 | 7, 8 | 882 | |||
XM_017006358.1 | XP_016861847.1 | 7, 8 | 882 | |||
GenBank mRNA | AK293752.1 | - | 1, 2 | 68 | ||
AK311008.1 | - | 7, 8 | 880 | |||
BC144563.1 | - | 7, 8 | 680 | |||
D26070.1 | - | 7, 8 | 788 | |||
L38019.2 | - | 7, 8 | 779 | |||
U23850.1 | - | 5, 6 | 532 |