Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003679.3 | NP_001003679.1 | 4 | 392 | ||
NM_001003680.3 | NP_001003680.1 | 4 | 392 | |||
NM_001198687.1 | NP_001185616.1 | 3 | 357 | |||
NM_001198688.1 | NP_001185617.1 | 3 | 357 | |||
NM_001198689.1 | NP_001185618.1 | 3 | 357 | |||
NM_002303.5 | NP_002294.2 | 4 | 392 | |||
GenBank mRNA | AK290208.1 | - | 4 | 357 | ||
BC031086.1 | - | 3 | 357 | |||
BC131779.1 | - | 4 | 324 | |||
U43168.1 | - | 4 | 400 | |||
U50748.1 | - | 2 | 207 | |||
U52912.1 | - | 3 | 303 | |||
U52913.1 | - | 3 | 303 | |||
U52914.1 | - | 3 | 303 | |||
U66495.1 | - | 4 | 400 | |||
U66496.1 | - | 4 | 400 | |||
U66497.1 | - | 3 | 333 |