Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291902.1 | NP_001278831.1 | 23 | 5130 | ||
NM_002335.3 | NP_002326.2 | 23 | 5108 | |||
XM_005273994.2 | XP_005274051.1 | 24 | 5217 | |||
XM_011545029.1 | XP_011543331.1 | 24 | 5158 | |||
XM_011545030.1 | XP_011543332.1 | 23 | 5044 | |||
XM_011545031.1 | XP_011543333.1 | 24 | 5174 | |||
XM_017017735.1 | XP_016873224.1 | 17 | 3438 | |||
XM_017017736.1 | XP_016873225.1 | 14 | 2821 | |||
GenBank mRNA | AF064548.1 | - | 23 | 5050 | ||
AF077820.1 | - | 23 | 5090 | |||
BC098397.1 | - | 23 | 5055 |