Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002226.4 | NP_002217.3 | 4, 5 | 1126 | ||
NM_145159.2 | NP_660142.1 | 4, 5 | 1126 | |||
GenBank mRNA | AF003521.1 | - | 4, 5 | 754 | ||
AF020201.1 | - | 4, 5 | 773 | |||
AF029778.1 | - | 4, 5 | 1126 | |||
AF029779.1 | - | 4, 5 | 1126 | |||
BC032053.1 | - | 4, 5 | 789 | |||
Y14330.1 | - | 4, 5 | 674 |