Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002644.3 | NP_002635.2 | 3 | 273 | ||
XM_011509629.1 | XP_011507931.1 | 3 | 184 | |||
GenBank mRNA | AF272149.2 | - | 3 | 269 | ||
AK225471.1 | - | 3 | 273 | |||
BC110494.2 | - | 3 | 170 | 40007650 | ||
BC110495.2 | - | 3 | 170 | 40007652 | ||
CR749533.1 | - | 4 | 346 | |||
S62403.1 | - | 3 | 245 | |||
X73079.1 | - | 3 | 269 |