Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002264.3 | NP_002255.3 | 3 | 392 | ||
NR_026698.1 | 3 | 392 | ||||
XM_005247437.3 | XP_005247494.1 | 3 | 274 | |||
XM_011512795.2 | XP_011511097.1 | 4 | 326 | |||
XM_011512797.2 | XP_011511099.1 | 2 | 209 | |||
XM_017006363.1 | XP_016861852.1 | 3 | 390 | |||
XM_017006364.1 | XP_016861853.1 | 3 | 269 | |||
GenBank mRNA | AK299030.1 | - | 2 | 278 | ||
AK312713.1 | - | 3 | 387 | |||
BC002374.2 | - | 3 | 381 | 2822859 | ||
BC003009.1 | - | 3 | 381 | 2822859 | ||
BC090864.1 | - | 3 | 363 | 6173605 | ||
BT006959.1 | - | 2 | 204 | |||
CR456743.1 | - | 2 | 204 | |||
DA308391.1 | - | 3 | 392 | |||
DN991078.1 | - | 3 | 234 | |||
S75295.1 | - | 3 | 250 | |||
U20620.1 | - | 1 | 57 |