Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206855.1 | NP_001193784.1 | 15 | 2456 | ||
NM_003227.3 | NP_003218.2 | 18 | 2876 | |||
XM_005250553.4 | XP_005250610.1 | 19 | 3182 | |||
XM_017012573.1 | XP_016868062.1 | 19 | 2984 | |||
GenBank mRNA | AF067864.1 | - | 18 | 2862 |