Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIA2 SIRNA Assays ›
Gene Symbol:
PPFIA2
Gene Name:
PTPRF interacting protein alpha 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 81257975 - 81759748 on Build GRCh38
UniGene ID:
Hs.506216
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
111325
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220473.2
NP_001207402.1
5
745
NM_001220474.2
NP_001207403.1
4
691
NM_001220475.2
NP_001207404.1
5
557
NM_001220476.2
NP_001207405.1
5
557
NM_001220477.2
NP_001207406.1
3
564
NM_001220478.2
NP_001207407.1
3
564
NM_001282536.1
NP_001269465.1
3
252
NM_003625.4
NP_003616.2
6
608
XM_006719670.3
XP_006719733.1
3
7800
XM_011538906.2
XP_011537208.1
4
685
XM_011538907.2
XP_011537209.1
4
685
XM_017020079.1
XP_016875568.1
6
592
XM_017020080.1
XP_016875569.1
5
739
XM_017020081.1
XP_016875570.1
5
739
XM_017020082.1
XP_016875571.1
6
592
XM_017020083.1
XP_016875572.1
5
739
XM_017020084.1
XP_016875573.1
5
739
XM_017020085.1
XP_016875574.1
5
739
XM_017020086.1
XP_016875575.1
6
592
XM_017020087.1
XP_016875576.1
5
726
XM_017020088.1
XP_016875577.1
5
972
XM_017020089.1
XP_016875578.1
4
685
XM_017020090.1
XP_016875579.1
6
592
XM_017020091.1
XP_016875580.1
5
739
XM_017020092.1
XP_016875581.1
5
739
XM_017020093.1
XP_016875582.1
5
739
XM_017020094.1
XP_016875583.1
6
592
XM_017020095.1
XP_016875584.1
5
739
XM_017020096.1
XP_016875585.1
5
739
XM_017020097.1
XP_016875586.1
6
592
XM_017020098.1
XP_016875587.1
6
592
XM_017020099.1
XP_016875588.1
4
685
XM_017020100.1
XP_016875589.1
5
739
XM_017020101.1
XP_016875590.1
6
592
XM_017020102.1
XP_016875591.1
5
726
XM_017020103.1
XP_016875592.1
5
739
XM_017020104.1
XP_016875593.1
5
739
XM_017020105.1
XP_016875594.1
5
739
XM_017020106.1
XP_016875595.1
5
739
XM_017020107.1
XP_016875596.1
6
592
XM_017020108.1
XP_016875597.1
6
592
XM_017020109.1
XP_016875598.1
5
739
XM_017020110.1
XP_016875599.1
5
972
XM_017020111.1
XP_016875600.1
5
739
XM_017020112.1
XP_016875601.1
3
564
XM_017020113.1
XP_016875602.1
3
564
XM_017020114.1
XP_016875603.1
3
564
XM_017020115.1
XP_016875604.1
3
564
XM_017020116.1
XP_016875605.1
3
564
XM_017020117.1
XP_016875606.1
3
374
XM_017020118.1
XP_016875607.1
4
539
XM_017020119.1
XP_016875608.1
3
564
XM_017020120.1
XP_016875609.1
3
252
GenBank mRNA
AB210009.1
-
5
743
AF034799.1
-
6
616
AK294505.1
-
3
374
AK297372.1
-
4
539
AK299853.1
-
3
513
AK302658.1
-
3
223
AK307524.1
-
6
592
BC104912.1
-
5
541
8143915
BC143485.1
-
5
541
9051993
DB083201.1
-
3
252
DC321269.1
-
3
533
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
neurotransmitter secretion
glutamate secretion
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0