Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL18R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282399.1 | NP_001269328.1 | 9 | 1052 | ||
NM_003855.3 | NP_003846.1 | 9 | 1242 | |||
XM_005264039.3 | XP_005264096.1 | 8 | 1121 | |||
XM_005264040.3 | XP_005264097.1 | 8 | 1124 | |||
XM_017005181.1 | XP_016860670.1 | 9 | 1181 | |||
XM_017005182.1 | XP_016860671.1 | 8 | 1463 | |||
XM_017005183.1 | XP_016860672.1 | 8 | 1461 | |||
XM_017005184.1 | XP_016860673.1 | 7 | 1295 | |||
GenBank mRNA | BC069575.1 | - | 8 | 1120 | 7262302 | |
BC093975.1 | - | 8 | 1120 | 7939820 | ||
BC093977.1 | - | 8 | 1120 | 7939822 | ||
BC143403.1 | - | 8 | 1117 | 9051910 | ||
BC143405.1 | - | 8 | 930 | |||
KM264374.1 | - | 5 | 511 | |||
KM264375.1 | - | 5 | 533 | |||
U43672.1 | - | 8 | 1120 |