Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSMR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168355.2 | NP_001161827.1 | 5 | 775 | ||
NM_001323504.1 | NP_001310433.1 | 5 | 895 | |||
NM_001323505.1 | NP_001310434.1 | 5 | 775 | |||
NM_001323506.1 | NP_001310435.1 | 5 | 775 | |||
NM_001323507.1 | NP_001310436.1 | 5 | 775 | |||
NM_003999.2 | NP_003990.1 | 5 | 895 | |||
XM_005248384.1 | XP_005248441.1 | 5 | 882 | |||
XM_005248386.2 | XP_005248443.1 | 5 | 882 | |||
XM_005248387.2 | XP_005248444.1 | 5 | 882 | |||
XM_011514161.2 | XP_011512463.1 | 5 | 882 | |||
XM_017010019.1 | XP_016865508.1 | 5 | 882 | |||
GenBank mRNA | BC010943.1 | - | 5 | 625 | ||
BC063468.1 | - | 5 | 547 | |||
BC125209.1 | - | 5 | 665 | 40123501 | ||
BC125210.1 | - | 5 | 665 | 40123502 | ||
U60805.1 | - | 5 | 860 |