Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDXR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258012.3 | NP_001244941.2 | 10 | 1297 | ||
NM_001258013.3 | NP_001244942.2 | 11 | 1261 | |||
NM_001258014.3 | NP_001244943.2 | 10 | 1144 | |||
NM_001258015.2 | NP_001244944.1 | 9 | 1048 | |||
NM_001258016.3 | NP_001244945.2 | 9 | 1113 | |||
NM_004110.5 | NP_004101.3 | 10 | 1186 | |||
NM_024417.4 | NP_077728.3 | 10 | 1168 | |||
NR_047576.2 | 10 | 1268 | ||||
XM_006721772.1 | XP_006721835.1 | 10 | 1116 | |||
XM_011524527.1 | XP_011522829.1 | 10 | 1134 | |||
XM_011524528.1 | XP_011522830.1 | 10 | 1223 | |||
XM_011524529.1 | XP_011522831.1 | 10 | 1226 | |||
XM_011524531.2 | XP_011522833.1 | 9 | 1119 | |||
XM_011524532.2 | XP_011522834.1 | 9 | 1126 | |||
XM_011524533.2 | XP_011522835.1 | 9 | 1094 | |||
GenBank mRNA | AK094120.1 | - | 10 | 1446 | ||
AK098648.1 | - | 10 | 1105 | |||
AK223530.1 | - | 10 | 1098 | |||
AK293208.1 | - | 11 | 1206 | |||
AK293210.1 | - | 10 | 1205 | |||
AK295307.1 | - | 10 | 1297 | |||
AK298908.1 | - | 9 | 991 | |||
AK301779.1 | - | 10 | 1081 | |||
AK301977.1 | - | 9 | 1113 | |||
BC002960.1 | - | 10 | 1107 | 3537008 | ||
BC018911.2 | - | 10 | 1114 | |||
BC034763.1 | - | 10 | 1769 | |||
BC063493.1 | - | 10 | 1106 | 5404814 | ||
BT006684.1 | - | 10 | 1081 | |||
J03826.1 | - | 10 | 1119 |