Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018054.2 | NP_001018064.1 | 8 | 1427 | ||
NM_004631.4 | NP_004622.2 | 8 | 1427 | |||
NM_017522.4 | NP_059992.3 | 7 | 1040 | |||
NM_033300.3 | NP_150643.2 | 6 | 917 | |||
XM_005271173.3 | XP_005271230.1 | 9 | 1376 | |||
XM_005271174.3 | XP_005271231.1 | 8 | 984 | |||
XM_005271175.3 | XP_005271232.1 | 7 | 945 | |||
XM_006710881.3 | XP_006710944.1 | 9 | 1376 | |||
XM_006710882.3 | XP_006710945.1 | 9 | 1376 | |||
XM_011542095.2 | XP_011540397.1 | 7 | 855 | |||
XM_011542096.2 | XP_011540398.1 | 9 | 1376 | |||
XM_017002266.1 | XP_016857755.1 | 9 | 1376 | |||
XM_017002267.1 | XP_016857756.1 | 7 | 821 | |||
XM_017002268.1 | XP_016857757.1 | 8 | 860 | |||
GenBank mRNA | AL119002.1 | - | 1 | 79 | ||
BC051836.1 | - | 8 | 1332 | 6143442 | ||
BE543079.1 | - | 4 | 256 | |||
D50678.1 | - | 8 | 1292 | |||
Z75190.1 | - | 7 | 803 |