Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005534.3 | NP_005525.2 | 3 | 963 | ||
XM_005260969.2 | XP_005261026.1 | 4 | 479 | |||
XM_011529553.1 | XP_011527855.1 | 4 | 405 | |||
XM_011529554.2 | XP_011527856.1 | 3 | 839 | |||
GenBank mRNA | AK292246.1 | - | 3 | 411 | ||
AK314005.1 | - | 3 | 424 | |||
BC003624.2 | - | 3 | 431 | 2967074 | ||
CB992729.1 | - | 3 | 473 | |||
EF173886.1 | - | 2 | 196 | |||
U05875.1 | - | 3 | 963 | |||
U05877.1 | - | 3 | 428 |