Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005608.2 | NP_005599.1 | 2 | 401 | ||
GenBank mRNA | AK312022.1 | - | 2 | 401 | ||
BC113859.1 | - | 2 | 400 | 40030438 | ||
BC114467.1 | - | 2 | 400 | 40030437 | ||
CR457429.1 | - | 1 | 338 | |||
X81422.1 | - | 2 | 401 |