Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278650.1 | NP_001265579.1 | 3 | 2452 | ||
NM_006057.2 | NP_006048.1 | 3 | 2604 | |||
NM_033170.2 | NP_149360.1 | 3 | 2471 | |||
NM_033171.2 | NP_149361.1 | 4 | 2713 | |||
NM_033172.2 | NP_149362.2 | 3 | 2553 | |||
XM_017028227.1 | XP_016883716.1 | 4 | 4029 | |||
XM_017028228.1 | XP_016883717.1 | 5 | 6653 | |||
XM_017028229.1 | XP_016883718.1 | 5 | 4191 | |||
XM_017028230.1 | XP_016883719.1 | 5 | 4206 | |||
XM_017028231.1 | XP_016883720.1 | 6 | 6762 | |||
XM_017028232.1 | XP_016883721.1 | 6 | 4249 | |||
XM_017028233.1 | XP_016883722.1 | 4 | 4026 | |||
XM_017028234.1 | XP_016883723.1 | 5 | 4188 | |||
XM_017028235.1 | XP_016883724.1 | 4 | 2629 | |||
XM_017028236.1 | XP_016883725.1 | 5 | 4587 | |||
XM_017028237.1 | XP_016883726.1 | 4 | 4702 | |||
XM_017028238.1 | XP_016883727.1 | 5 | 6520 | |||
XM_017028239.1 | XP_016883728.1 | 6 | 6629 | |||
XM_017028240.1 | XP_016883729.1 | 3 | 3798 | |||
XM_017028241.1 | XP_016883730.1 | 3 | 3795 | |||
GenBank mRNA | AB020337.1 | - | 3 | 2597 | ||
AY372061.1 | - | 5 | 2756 |