Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002157.2 | NP_002148.1 | 2, 3 | 611 | ||
GenBank mRNA | AK312104.1 | - | 2, 3 | 207 | ||
BC023518.2 | - | 2, 3 | 285 | 4304224 | ||
BG188453.1 | - | 1, 2 | 172 | |||
CN264788.1 | - | 2, 3 | 616 | |||
CR407688.1 | - | 1, 2 | 171 | |||
U07550.1 | - | 2, 3 | 212 | |||
X75821.1 | - | 1, 2 | 171 |