Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195007.1 | NP_001181936.1 | 6, 7 | 448 | ||
NM_006112.3 | NP_006103.1 | 6, 7 | 448 | |||
NM_203456.2 | NP_982281.1 | 6, 7 | 448 | |||
NR_036543.1 | 6, 7 | 440 | ||||
XM_011540501.1 | XP_011538803.1 | 6, 7 | 550 | |||
XM_017000051.1 | XP_016855540.1 | 6, 7 | 399 | |||
XM_017000052.1 | XP_016855541.1 | 6, 7 | 550 | |||
GenBank mRNA | AF042385.1 | - | 6, 7 | 452 | ||
AF042386.1 | - | 6, 7 | 452 | |||
AF104012.1 | - | 6, 7 | 403 | |||
AF104013.1 | - | 6, 7 | 403 | |||
AK094660.1 | - | 5, 6 | 1675 | |||
AK130628.1 | - | 6, 7 | 401 | |||
AK293094.1 | - | 6, 7 | 399 | |||
AK313666.1 | - | 6, 7 | 416 | |||
AL833576.1 | - | 6, 7 | 508 | |||
BC004898.2 | - | 6, 7 | 398 | 3545997 | ||
BC008451.1 | - | 6, 7 | 411 | 4134572 | ||
BC107736.1 | - | 6, 7 | 407 | 4809032 | ||
BU160972.1 | - | 6, 7 | 377 | |||
BU179717.1 | - | 435 | ||||
BU196539.1 | - | 6, 7 | 435 | |||
DQ160195.1 | - | 6, 7 | 392 |