Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006437.3 | NP_006428.2 | 14, 15 | 1896 | ||
XM_011534931.1 | XP_011533233.1 | 13, 14 | 1946 | |||
XM_011534932.2 | XP_011533234.1 | 14, 15 | 2113 | |||
GenBank mRNA | AF057160.1 | - | 14, 15 | 1842 | ||
AF158255.1 | - | 14, 15 | 1896 | |||
AK304399.1 | - | 14, 15 | 1864 | |||
D79999.2 | - | 13, 14 | 1795 |