Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319075.1 | NP_001306004.1 | 2 | 1953 | ||
NM_006577.5 | NP_006568.2 | 2 | 2087 | |||
GenBank mRNA | AF288208.1 | - | 3 | 2034 | ||
AF288209.1 | - | 2 | 1836 | |||
AK002009.1 | - | 1 | 1782 | |||
BC030579.2 | - | 2 | 1956 | 4828158 | ||
BC047933.1 | - | 2 | 2086 | 5767816 |