Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006581.3 | NP_006572.2 | 3 | 984 | ||
XM_011535383.2 | XP_011533685.1 | 3 | 6721 | |||
XM_011535385.2 | XP_011533687.1 | 4 | 6838 | |||
XM_017010188.1 | XP_016865677.1 | 4 | 3985 | |||
XM_017010190.1 | XP_016865679.1 | 4 | 911 | |||
GenBank mRNA | AB023021.1 | - | 3 | 937 | ||
AJ238701.1 | - | 2 | 747 | |||
AK314966.1 | - | 3 | 979 | |||
AY800264.1 | - | 1 | 643 | |||
BC036101.1 | - | 3 | 961 | 5286309 |