Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012405.3 | NP_036537.1 | 3 | 467 | ||
XM_011541140.2 | XP_011539442.1 | 2 | 167 | |||
GenBank mRNA | AF064084.1 | - | 3 | 437 | ||
AK092890.1 | - | 4 | 408 | |||
BC017037.1 | - | 4 | 562 | |||
BC028168.1 | - | 3 | 423 | 5245125 | ||
CR541671.1 | - | 3 | 394 | |||
CR541711.1 | - | 3 | 394 |