Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013339.3 | NP_037471.2 | 11 | 1283 | ||
GenBank mRNA | AF063604.2 | - | 11 | 1238 | ||
AF102851.1 | - | 11 | 1216 | |||
AK022700.1 | - | 11 | 1221 | |||
BC001253.1 | - | 11 | 1215 | 3452687 | ||
CB215801.1 | - | 3 | 209 |