Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNMT3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013369.3 | NP_037501.2 | 3, 4 | 627 | ||
NM_175867.2 | NP_787063.1 | 3, 4 | 627 | |||
GenBank mRNA | AF194032.1 | - | 3, 4 | 627 | ||
BC002560.2 | - | 3, 4 | 301 | 3138514 |