Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017417.1 | NP_059113.1 | 2 | 369 | ||
GenBank mRNA | AJ271385.1 | - | 2 | 369 | ||
BC140888.1 | - | 2 | 335 | 9021756 | ||
BC140889.1 | - | 2 | 335 | 9021757 |