Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017584.5 | NP_060054.4 | 6, 7 | 626 | ||
XM_005261925.3 | XP_005261982.1 | 5, 6 | 493 | |||
GenBank mRNA | AF197129.1 | - | 5, 6 | 512 | ||
AY064416.1 | - | 6, 7 | 539 | |||
AY738258.1 | - | 5, 6 | 512 | |||
BC012025.1 | - | 6, 7 | 658 | 4454512 | ||
BC064395.1 | - | 6, 7 | 534 | |||
BC073848.1 | - | 6, 7 | 546 | 5186354 | ||
CR456478.1 | - | 6, 7 | 571 |