Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317947.1 | NP_001304876.1 | 3 | 747 | ||
NM_018146.3 | NP_060616.1 | 4 | 992 | |||
GenBank mRNA | AF177344.2 | - | 4 | 1048 | ||
AK001443.1 | - | 4 | 968 | |||
AK222900.1 | - | 4 | 968 | |||
AK225128.1 | - | 4 | 949 | |||
BC011550.1 | - | 4 | 954 | 4620063 | ||
BC050614.1 | - | 4 | 950 | 5248052 |