Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318865.1 | NP_001305794.1 | 6 | 796 | ||
NM_018960.5 | NP_061833.1 | 6 | 853 | |||
NR_134899.1 | 5 | 710 | ||||
GenBank mRNA | AF101477.1 | - | 6 | 849 | ||
BC032627.1 | - | 6 | 845 | 5229272 | ||
FJ224320.1 | - | 6 | 849 | |||
KU178603.1 | - | 6 | 839 | |||
KU178604.1 | - | 6 | 782 | |||
KU178605.1 | - | 5 | 696 |
Gene Symbol: CNPY3-GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318856.1 | NP_001305785.1 | 6 | 1225 | ||
NM_001318857.1 | NP_001305786.1 | 5 | 1097 | |||
NM_001318858.1 | NP_001305787.1 | 5 | 989 | |||
NR_134890.1 | 5 | 1051 | ||||
NR_134891.1 | 4 | 954 | ||||
NR_134892.1 | 5 | 1082 |