Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034023.1 | NP_001029195.1 | 5 | 632 | ||
NM_001034024.1 | NP_001029196.1 | 5 | 592 | |||
NM_019100.4 | NP_061973.1 | 4 | 813 | |||
XM_005271037.4 | XP_005271094.1 | 4 | 803 | |||
XM_005271039.1 | XP_005271096.1 | 5 | 703 | |||
XM_005271040.1 | XP_005271097.1 | 5 | 580 | |||
XM_006710771.3 | XP_006710834.1 | 3 | 482 | |||
XM_011541784.2 | XP_011540086.1 | 5 | 637 | |||
XM_011541785.2 | XP_011540087.1 | 5 | 657 | |||
XM_017001807.1 | XP_016857296.1 | 5 | 688 | |||
XM_017001808.1 | XP_016857297.1 | 5 | 691 | |||
GenBank mRNA | AB037846.1 | - | 5 | 597 | ||
AF265228.1 | - | 4 | 512 | |||
AK021605.1 | - | 5 | 576 | |||
AK054954.1 | - | 1 | 1430 | |||
AK092898.1 | - | 1 | 3597 | |||
AK096252.1 | - | 1 | 2200 | |||
AK289366.1 | - | 5 | 615 | |||
AK293598.1 | - | 5 | 739 | |||
AK298793.1 | - | 5 | 575 | |||
AL136657.1 | - | 5 | 594 | |||
AL137200.1 | - | 5 | 596 | |||
BC002855.2 | - | 5 | 581 | 3635724 | ||
BC008053.2 | - | 4 | 775 | 3834056 |