Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGPAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020133.2 | NP_064518.1 | 2 | 308 | ||
XM_005267053.3 | XP_005267110.1 | 2 | 294 | |||
XM_006715514.3 | XP_006715577.1 | 2 | 294 | |||
XM_017011059.1 | XP_016866548.1 | 2 | 341 | |||
GenBank mRNA | AF156776.1 | - | 2 | 238 | ||
AJ227890.1 | - | 1 | 104 | |||
AJ420473.1 | - | 2 | 280 | |||
AK074208.1 | - | 2 | 294 | |||
AK295025.1 | - | 2 | 255 | |||
AK295548.1 | - | 2 | 252 | |||
AK295826.1 | - | 2 | 231 | |||
AK299721.1 | - | 2 | 278 | |||
AY358506.1 | - | 2 | 248 | |||
BC020209.1 | - | 2 | 240 | 4850399 | ||
BQ072907.1 | - | 308 | ||||
CR627395.1 | - | 2 | 259 |