Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KISS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002256.3 | NP_002247.3 | 3 | 695 | ||
GenBank mRNA | AF218017.1 | - | 3 | 619 | ||
AK291695.1 | - | 3 | 694 | |||
BC022819.1 | - | 3 | 690 | 5443510 | ||
U43527.1 | - | 3 | 752 |