Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017403.1 | NP_001017403.1 | 3 | 308 | ||
NM_001017404.1 | NP_001017404.1 | 3 | 179 | |||
NM_021636.2 | NP_067649.2 | 3 | 195 | |||
XM_005245404.3 | XP_005245461.1 | 3 | 252 | |||
XM_011509838.2 | XP_011508140.1 | 3 | 253 | |||
XM_011509839.2 | XP_011508141.1 | 3 | 254 | |||
XM_011509840.2 | XP_011508142.1 | 3 | 253 | |||
XM_017001996.1 | XP_016857485.1 | 3 | 254 | |||
GenBank mRNA | AB049405.1 | - | 3 | 193 | ||
AF190501.1 | - | 3 | 179 | |||
AY358119.1 | - | 3 | 236 | |||
BC047905.1 | - | 3 | 178 | 5212508 |