Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002267.3 | NP_002258.2 | 6, 7 | 797 | ||
XM_017020561.1 | XP_016876050.1 | 6, 7 | 480 | |||
GenBank mRNA | AF005263.1 | - | 6, 7 | 394 | ||
AF034756.1 | - | 6, 7 | 384 | |||
AK223480.1 | - | 6, 7 | 419 | |||
AK290528.1 | - | 6, 7 | 457 | |||
AK291000.1 | - | 6, 7 | 421 | |||
BC017355.2 | - | 6, 7 | 406 | 5020312 | ||
BC024202.2 | - | 6, 7 | 406 | 3531069 | ||
BC035090.2 | - | 6, 7 | 798 | 5261465 | ||
D89618.1 | - | 6, 7 | 473 | |||
Y12394.1 | - | 6, 7 | 397 |