Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173353.3 | NP_775489.2 | 2 | 307 | ||
GenBank mRNA | AK094614.1 | - | 2 | 176 | ||
AY098914.1 | - | 2 | 307 | |||
BC114442.1 | - | 2 | 262 | 40083878 |