Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST3GAL3 SIRNA Assays ›
Gene Symbol:
ST3GAL3
Gene Name:
ST3 beta-galactoside alpha-2,3-sialyltransferase 3
Gene Aliases:
EIEE15, MRT12, SIAT6, ST3GALII, ST3GalIII, ST3N
Chromosome Location:
Chr.1: 43707497 - 43931165 on Build GRCh38
UniGene ID:
Hs.597915
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
112418
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST3GAL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270459.1
NP_001257388.1
5
426
NM_001270461.1
NP_001257390.1
4
378
NM_001270463.1
NP_001257392.1
4
423
NM_001270464.1
NP_001257393.1
4
378
NM_001270465.1
NP_001257394.1
4
423
NM_001270466.1
NP_001257395.1
4
378
NM_006279.3
NP_006270.1
5
426
NM_174963.3
NP_777623.2
6
633
NM_174964.2
NP_777624.1
5
471
NM_174965.2
NP_777625.1
5
471
NM_174966.2
NP_777626.1
5
426
NM_174967.2
NP_777627.1
5
426
NM_174968.3
NP_777628.2
6
588
NM_174969.2
NP_777629.1
4
378
NM_174970.2
NP_777630.1
4
378
NM_174971.3
NP_777631.2
5
540
NR_073016.1
5
426
NR_073017.1
3
335
NR_073018.1
5
426
NR_073020.1
4
378
NR_073021.1
5
426
NR_073023.1
5
426
XM_006710825.3
XP_006710888.1
5
462
XM_006710827.3
XP_006710890.1
4
380
XM_006710828.3
XP_006710891.1
5
428
XM_011541973.2
XP_011540275.1
6
635
XM_011541974.2
XP_011540276.1
6
590
XM_011541976.2
XP_011540278.1
5
542
XM_011541977.2
XP_011540279.1
5
473
XM_011541980.2
XP_011540282.1
2
693
XM_011541981.2
XP_011540283.1
6
635
XM_011541983.2
XP_011540285.1
6
737
XM_011541984.2
XP_011540286.1
6
635
XM_011541985.2
XP_011540287.1
6
635
XM_011541986.2
XP_011540288.1
6
635
XM_011541987.2
XP_011540289.1
5
428
XM_017002109.1
XP_016857598.1
6
698
XM_017002110.1
XP_016857599.1
6
635
XM_017002111.1
XP_016857600.1
6
691
XM_017002112.1
XP_016857601.1
6
687
XM_017002113.1
XP_016857602.1
5
506
XM_017002114.1
XP_016857603.1
5
529
XM_017002115.1
XP_016857604.1
4
380
XM_017002116.1
XP_016857605.1
5
525
XM_017002117.1
XP_016857606.1
6
590
XM_017002118.1
XP_016857607.1
5
473
XM_017002119.1
XP_016857608.1
5
426
XM_017002120.1
XP_016857609.1
5
425
XM_017002121.1
XP_016857610.1
3
218
XM_017002122.1
XP_016857611.1
5
529
GenBank mRNA
AF425851.1
-
4
280
AF425852.1
-
4
280
AF425853.1
-
5
442
AF425854.1
-
4
235
AF425856.1
-
4
235
AF425857.1
-
4
235
AF425858.1
-
5
397
AF425859.1
-
3
187
AF425860.1
-
3
187
AF425861.1
-
4
349
AF425862.1
-
4
235
AF425863.1
-
3
232
AF425864.1
-
4
235
AF425865.1
-
4
235
AF425866.1
-
4
235
AF425867.1
-
4
235
AF425868.1
-
2
144
AK301829.1
-
5
412
AK310265.1
-
6
704
AY051143.1
-
4
363
AY167992.1
-
4
235
AY167993.1
-
3
187
AY167994.1
-
3
232
AY167995.1
-
3
187
AY167996.1
-
3
187
AY167998.1
-
3
187
BC050380.1
-
5
410
5769575
L23768.1
-
4
235
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001285521, NM_009176, NM_001285520, NM_001161774, BC006710, and others.
Panther Classification:
Molecular Function -
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
ganglioside biosynthetic process
oligosaccharide metabolic process
O-glycan processing
keratan sulfate biosynthetic process
protein N-linked glycosylation via asparagine
sialylation
beta-galactoside (CMP) alpha-2,3-sialyltransferase activity
N-acetyllactosaminide alpha-2,3-sialyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0