Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004605.2 | NP_004596.2 | 3, 4 | 686 | ||
NM_177973.1 | NP_814444.1 | 4, 5 | 633 | |||
GenBank mRNA | BC034694.1 | - | 4, 5 | 622 | 4745903 | |
U92314.1 | - | 3, 4 | 882 | |||
U92315.1 | - | 4, 5 | 633 |