Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004480.4 | NP_004471.4 | 9 | 2682 | ||
NM_178155.2 | NP_835368.1 | 10 | 3112 | |||
NM_178156.2 | NP_835369.1 | 9 | 3014 | |||
NR_038167.1 | 8 | 2688 | ||||
NR_038170.1 | 10 | 2195 | ||||
XM_011536614.2 | XP_011534916.1 | 5 | 867 | |||
XM_017021136.1 | XP_016876625.1 | 11 | 2297 | |||
XM_017021137.1 | XP_016876626.1 | 11 | 1941 | |||
XM_017021138.1 | XP_016876627.1 | 10 | 1808 | |||
XM_017021139.1 | XP_016876628.1 | 10 | 2199 | |||
XM_017021140.1 | XP_016876629.1 | 9 | 4149 | |||
GenBank mRNA | AB049740.2 | - | 1254 | |||
AF052088.1 | - | 4 | 751 | |||
AJ514324.1 | - | 2 | 192 | |||
AJ514325.1 | - | 2 | 192 | |||
AJ536053.1 | - | 10 | 2580 | |||
AJ536054.1 | - | 10 | 2580 | |||
AJ536055.1 | - | 9 | 2482 | |||
AJ536056.1 | - | 10 | 3110 | |||
AJ539535.1 | - | 9 | 1710 | |||
AJ539536.1 | - | 9 | 1710 | |||
AK292413.1 | - | 10 | 1833 | |||
AK294242.1 | - | 9 | 1400 | |||
BC025385.2 | - | 10 | 1589 | |||
BC093889.1 | - | 2 | 263 | |||
BC101816.1 | - | 2 | 263 | |||
BC142958.1 | - | 8 | 1415 | 8860361 | ||
D89289.1 | - | 8 | 1527 | |||
Y17976.1 | - | 10 | 1813 | |||
Y17977.1 | - | 10 | 1813 | |||
Y17978.1 | - | 10 | 2101 | |||
Y17979.1 | - | 10 | 2101 |