Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOGAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287147.1 | NP_001274076.1 | 3 | 444 | ||
NM_178176.3 | NP_835470.1 | 3 | 444 | |||
XM_005250309.3 | XP_005250366.1 | 3 | 554 | |||
XM_011516126.2 | XP_011514428.1 | 2 | 446 | |||
XM_011516127.2 | XP_011514429.1 | 2 | 169 | |||
GenBank mRNA | AY229854.1 | - | 3 | 444 | ||
AY358200.1 | - | 3 | 421 | |||
BC100953.1 | - | 3 | 335 | 40004561 | ||
BC100954.1 | - | 3 | 335 | 40004562 | ||
BC100955.1 | - | 3 | 338 | 40004564 |