Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198066.3 | NP_932332.1 | 3 | 463 | ||
XM_005268012.2 | XP_005268069.1 | 4 | 497 | |||
XM_006720238.3 | XP_006720301.1 | 3 | 596 | |||
GenBank mRNA | AK090577.1 | - | 3 | 396 | ||
BC012179.1 | - | 3 | 265 | 4661046 | ||
BG717628.1 | - | 4 | 505 |