Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | J02703.1 | - | 12 | 1975 | ||
M20311.1 | - | 12 | 1971 | |||
U95204.1 | - | 12 | 1955 |