Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000284.3 | NP_000275.1 | 10 | 1098 | ||
NM_001173454.1 | NP_001166925.1 | 11 | 1212 | |||
NM_001173455.1 | NP_001166926.1 | 10 | 1119 | |||
NM_001173456.1 | NP_001166927.1 | 9 | 1005 | |||
XM_017029574.1 | XP_016885063.1 | 10 | 1106 | |||
GenBank mRNA | AK129887.1 | - | 10 | 1033 | ||
AK222740.1 | - | 10 | 1029 | |||
AK222988.1 | - | 10 | 1063 | |||
AK293250.1 | - | 11 | 1177 | |||
AK296341.1 | - | 10 | 1085 | |||
AK296457.1 | - | 9 | 919 | |||
AK312263.1 | - | 10 | 1064 | |||
BC002406.1 | - | 10 | 1010 | 2961286 | ||
EF576990.1 | - | 7 | 567 | |||
EF590116.1 | - | 4 | 748 | |||
EF590117.1 | - | 1130 | ||||
J03503.1 | - | 10 | 1025 | |||
J03575.1 | - | 10 | 998 | |||
L13318.1 | - | 10 | 1058 | |||
L48690.1 | - | 10 | 1058 | |||
M24848.1 | - | 10 | 996 | |||
X52709.1 | - | 10 | 1007 | |||
X52710.1 | - | 10 | 1009 |