Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000420.2 | NP_000411.1 | 13, 14 | 1706 | ||
XM_005249993.2 | XP_005250050.1 | 13, 14 | 1557 | |||
XM_005249994.3 | XP_005250051.1 | 7, 8 | 877 | |||
GenBank mRNA | AK314831.1 | - | 13, 14 | 1607 | ||
BC003135.1 | - | 13, 14 | 1706 | 2906155 | ||
BC050639.1 | - | 13, 14 | 1601 | 5742350 | ||
FN998902.1 | - | 12, 13 | 1496 | |||
HE572750.1 | - | 12, 13 | 1496 | |||
M64934.2 | - | 13, 14 | 1616 |