Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCNN1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038.5 | NP_001029.1 | 13 | 3176 | ||
NM_001159575.1 | NP_001153047.1 | 13 | 3047 | |||
NM_001159576.1 | NP_001153048.1 | 12 | 3328 | |||
GenBank mRNA | AK025536.1 | - | 8 | 1900 | ||
AK172792.1 | - | 12 | 3331 | |||
BC006526.2 | - | 12 | 3014 | |||
BC062613.1 | - | 12 | 2943 | 5186683 | ||
X76180.1 | - | 13 | 2986 |