Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128928.1 | NP_001122400.1 | 7 | 2026 | ||
NM_002194.3 | NP_002185.1 | 6 | 1882 | |||
XM_005246532.1 | XP_005246589.1 | 6 | 1663 | |||
GenBank mRNA | AK093560.1 | - | 8 | 1984 | ||
BC015496.1 | - | 5 | 1392 | 3845956 | ||
BC106006.1 | - | 6 | 1426 | 4472604 | ||
L08488.1 | - | 6 | 1652 |