Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELANE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001972.3 | NP_001963.1 | 5 | 755 | ||
XM_011527775.1 | XP_011526077.1 | 6 | 1432 | |||
XM_011527776.1 | XP_011526078.1 | 6 | 771 | |||
GenBank mRNA | BC074816.2 | - | 5 | 672 | 30915534 | |
BC074817.2 | - | 5 | 672 | 30915320 | ||
D00187.1 | - | 4 | 545 | |||
J03545.1 | - | 3 | 413 | |||
M27783.1 | - | 2 | 269 | |||
M34379.1 | - | 5 | 673 | |||
X05875.1 | - | 5 | 673 |