Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002205.4 | NP_002196.4 | 30 | 4191 | ||
GenBank mRNA | BC008786.2 | - | 30 | 4176 | 3629647 | |
BX647780.1 | - | 17 | 4937 | |||
M13918.2 | - | 30 | 4075 | |||
X06256.1 | - | 30 | 4138 |