Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001118885.1 | NP_001112357.1 | 3 | 452 | ||
NM_001118886.1 | NP_001112358.1 | 2 | 880 | |||
NM_001171942.1 | NP_001165413.1 | 4 | 659 | |||
NM_002063.3 | NP_002054.1 | 2 | 880 | |||
XM_006724487.3 | XP_006724550.1 | 3 | 263 | |||
XM_011545495.2 | XP_011543797.1 | 4 | 937 | |||
XM_011545496.2 | XP_011543798.1 | 4 | 647 | |||
XM_017029427.1 | XP_016884916.1 | 3 | 424 | |||
XM_017029428.1 | XP_016884917.1 | 2 | 230 | |||
XM_017029429.1 | XP_016884918.1 | 3 | 775 | |||
GenBank mRNA | AK289561.1 | - | 2 | 407 | ||
AK297268.1 | - | 2 | 371 | |||
AK297304.1 | - | 4 | 596 | |||
AK312591.1 | - | 3 | 452 | |||
AY437083.1 | - | 2 | 120 | |||
AY437084.1 | - | 2 | 120 | |||
BC032864.2 | - | 2 | 650 | 5263356 | ||
CR450343.1 | - | 2 | 120 | |||
X52008.1 | - | 2 | 507 |