Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002301.4 | NP_002292.1 | 4 | 508 | ||
NM_017448.3 | NP_059144.1 | 4 | 454 | |||
XM_017017721.1 | XP_016873210.1 | 3 | 398 | |||
XM_017017722.1 | XP_016873211.1 | 3 | 319 | |||
GenBank mRNA | AF401094.1 | - | 3 | 282 | ||
AF401096.1 | - | 3 | 282 | |||
AF401097.1 | - | 4 | 400 | |||
AK058181.1 | - | 4 | 430 | |||
AY286300.1 | - | 4 | 430 | |||
BC019249.2 | - | 4 | 461 | 4839028 | ||
BC064388.1 | - | 4 | 477 | 5744600 | ||
BC090043.1 | - | 4 | 406 | 30387780 | ||
BG717645.1 | - | 4 | 454 | |||
GU479374.1 | - | 3 | 346 | |||
HQ386000.1 | - | 4 | 382 | |||
HQ386001.1 | - | 4 | 436 | |||
HQ386004.1 | - | 3 | 342 | |||
J02938.1 | - | 4 | 408 | |||
U13680.1 | - | 4 | 483 |