Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003941.2 | NP_001003941.1 | 2 | 255 | ||
NM_001165036.1 | NP_001158508.1 | 2 | 255 | |||
NM_002541.3 | NP_002532.2 | 2 | 255 | |||
XM_005249759.4 | XP_005249816.1 | 2 | 187 | |||
XM_005249761.3 | XP_005249818.1 | 2 | 205 | |||
XM_011515408.2 | XP_011513710.1 | 3 | 274 | |||
GenBank mRNA | AK296400.1 | - | 2 | 206 | ||
AK304439.1 | - | 2 | 186 | |||
AK314179.1 | - | 2 | 177 | |||
AU140451.1 | - | 2 | 209 | |||
BC004964.1 | - | 1 | 138 | 3528538 | ||
BC009580.1 | - | 2 | 189 | |||
BC014617.2 | - | 1 | 138 | 3010095 | ||
D10523.1 | - | 2 | 153 | |||
DC401267.1 | - | 2 | 255 |