Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002787.4 | NP_002778.1 | 6 | 516 | ||
GenBank mRNA | AK222976.1 | - | 6 | 484 | ||
AK290654.1 | - | 6 | 491 | |||
AU120930.1 | - | 6 | 491 | |||
BC002900.2 | - | 5 | 442 | |||
BC047697.1 | - | 6 | 470 | 5786518 | ||
BX641097.1 | - | 5 | 407 | |||
CB990982.1 | - | 6 | 559 | |||
CR450317.1 | - | 6 | 468 | |||
D00760.1 | - | 6 | 468 |