Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199771.1 | NP_001186700.1 | 5 | 1142 | ||
NM_002905.3 | NP_002896.2 | 5 | 1137 | |||
GenBank mRNA | BC028298.1 | - | 5 | 1096 | 4942311 | |
U43559.1 | - | 5 | 1030 | |||
U89717.1 | - | 5 | 1067 |
Gene Symbol: BLOC1S1-RDH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037658.1 | 6 | 1013 | |||
GenBank mRNA | AK096656.1 | - | 4 | 3204 |